Gene Transcripts

Gene Symbol
Alox12b
Entrez Gene ID
287425
Gene Name
arachidonate 12-lipoxygenase, 12R type
Gene Aliases
Chromosome Location
Chr. 10: 55744646 - 55756524 on Build Rnor_6.0
UniGene ID
Rn.160588
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039377.1 NP_001034466.1 11 1622
GenBank mRNA AY903231.1 - 11 1622
AY903232.1 - 11 1622
AY903233.1 - 11 1622

View Full Product Details