Gene Transcripts

Gene Symbol
RGD1310352
Entrez Gene ID
303122
Gene Name
similar to HTGN29 protein; keratinocytes associated transmembrane protein 2
Gene Aliases
Chromosome Location
Chr. 10: 37757134 - 37769781 on Build Rnor_6.0
UniGene ID
Rn.106999
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106999.1 NP_001100469.1 2 655
XM_006246261.2 XP_006246323.1 2 667
GenBank mRNA BC079420.1 - 2 624

View Full Product Details