Gene Transcripts
- Gene Symbol
- Ikbkg
- Entrez Gene ID
- 309295
- Gene Name
- inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma
- Gene Aliases
- Nemo
- Chromosome Location
- Chr. X: 156254187 - 156280070 on Build Rnor_6.0
- UniGene ID
- Rn.214715
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_199103.1 |
NP_954534.1
|
3, 4 |
522 |
|
XM_006229580.2 |
XP_006229642.1
|
5, 6 |
1770 |
|
XM_006229584.2 |
XP_006229646.1
|
5, 6 |
756 |
|
XM_006229585.2 |
XP_006229647.1
|
4, 5 |
716 |
|
XM_008773600.1 |
XP_008771822.1
|
4, 5 |
4830 |
|
XM_008773601.1 |
XP_008771823.1
|
5, 6 |
1773 |
|
XM_008773602.1 |
XP_008771824.1
|
5, 6 |
1787 |
|
XM_008773603.1 |
XP_008771825.1
|
5, 6 |
1778 |
|
XM_008773604.1 |
XP_008771826.1
|
4, 5 |
4827 |
|
XM_008773605.1 |
XP_008771827.1
|
4, 5 |
4827 |
|
XM_008773606.1 |
XP_008771828.1
|
4, 5 |
4824 |
|
XM_008773607.1 |
XP_008771829.1
|
5, 6 |
4870 |
|
XM_008773608.1 |
XP_008771830.1
|
5, 6 |
760 |
|
XM_008773609.1 |
XP_008771831.1
|
5, 6 |
968 |
|
XM_008773610.1 |
XP_008771832.1
|
4, 5 |
720 |
|
GenBank mRNA |
AY392762.1 |
- |
3, 4 |
522 |
|
View Full Product Details