Gene Transcripts

Gene Symbol
ASMTL
Entrez Gene ID
8623
Gene Name
acetylserotonin O-methyltransferase-like
Gene Aliases
ASMTLX, ASMTLY, ASTML
Chromosome Location
Chr. X: 1403139 - 1453794 on Build GRCh38
UniGene ID
Hs.533514
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001173473.1 NP_001166944.1 6 488
NM_001173474.1 NP_001166945.1 5 525
NM_004192.3 NP_004183.2 6 573
XM_005274434.3 XP_005274491.1 7 679
XM_011546185.2 XP_011544487.1 4 321
XM_017029919.1 XP_016885408.1 5 369
GenBank mRNA AK090498.1 - 5 477
AK225770.1 - 6 495
AK297805.1 - 5 406
AK299231.1 - 6 491
AK301844.1 - 6 482
BC002508.2 - 6 459 3051194
BC010089.2 - 6 457 2988425
BM129217.1 - 1 22
DA301876.1 - 6 528
DA904467.1 - 6 490

View Full Product Details