Gene Transcripts

Gene Symbol
Olr1509
Entrez Gene ID
363642
Gene Name
olfactory receptor 1509
Gene Aliases
Chromosome Location
Chr. 10: 60941416 - 60942351 on Build Rnor_6.0
UniGene ID
Rn.142079
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000528.1 NP_001000528.1 1 586

View Full Product Details