Gene Transcripts

Gene Symbol
LOC499544
Entrez Gene ID
499544
Gene Name
LRRGT00154
Gene Aliases
Chromosome Location
Chr. 2: 61991742 - 62018698 on Build Rnor_6.0
UniGene ID
Rn.161787
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047944.1 NP_001041409.1 3 146
GenBank mRNA AY539905.1 - 3 146

View Full Product Details