Gene Transcripts

Gene Symbol
LOC689713
Entrez Gene ID
689713
Gene Name
LRRGT00175
Gene Aliases
Chromosome Location
Chr. 16: 84465656 - 84495583 on Build Rnor_6.0
UniGene ID
Rn.123858
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047978.1 NP_001041443.1 6 493
GenBank mRNA AY539926.1 - 6 493

View Full Product Details