Gene Transcripts
- Gene Symbol
- LOC100630923
- Entrez Gene ID
- 100630923
- Gene Name
- LOC100289561-PRKRIP1 readthrough
- Gene Aliases
- Chromosome Location
- Chr. 7: 102363861 - 102426682 on Build GRCh38
- UniGene ID
- Hs.406395
- Species
- Homo sapiens
- Gene Symbol
- LOC100289561
- Entrez Gene ID
- 100289561
- Gene Name
- uncharacterized LOC100289561
- Gene Aliases
- Chromosome Location
- Chr. 7: 102363861 - 102380633 on Build GRCh38
- UniGene ID
- Hs.406395
- Species
- Homo sapiens
- Gene Symbol
- PMS2P3
- Entrez Gene ID
- 5387
- Gene Name
- PMS1 homolog 2, mismatch repair system component pseudogene 3
- Gene Aliases
- PMS2L3, PMS2L9, PMS5, PMSR3
- Chromosome Location
- Chr. 7: 75507747 - 75528123 on Build GRCh38
- UniGene ID
- Hs.659871
- Species
- Homo sapiens
View Full Product Details