Gene Transcripts

Gene Symbol
ANP32B
Entrez Gene ID
10541
Gene Name
acidic nuclear phosphoprotein 32 family member B
Gene Aliases
APRIL, PHAPI2, SSP29
Chromosome Location
Chr. 9: 97983207 - 98015943 on Build GRCh38
UniGene ID
Hs.730654
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006401.2 NP_006392.1 3, 4 671
GenBank mRNA AK223454.1 - 3, 4 538
AK313733.1 - 3, 4 526
BC013003.2 - 3, 4 503 3010189
BC019658.1 - 3, 4 510 4478860
U70439.1 - 3, 4 532
Y07569.1 - 3, 4 377
Y07570.1 - 3, 4 506
Y07969.1 - 3, 4 546

View Full Product Details