Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001031735.2 | NP_001026905.2 | 1 | 252 | |
NM_001039846.1 | NP_001034935.1 | 1 | 252 | ||
XM_005259480.3 | XP_005259537.2 | 1 | 273 | ||
GenBank mRNA | AK294083.1 | - | 1 | 123 | |
AK301957.1 | - | 1 | 206 | ||
AY358444.1 | - | 1 | 252 | ||
AY753639.1 | - | 1 | 211 | ||
BC010443.1 | - | 1 | 191 | ||
BC014609.2 | - | 1 | 206 | 4825119 | |
BC040964.1 | - | 1 | 193 | ||
BI827974.1 | - | 1 | 109 | ||
BX378854.2 | - | 1 | 252 | ||
DQ196309.1 | - | 1 | 239 | ||
DQ196310.1 | - | 1 | 239 | ||
DQ424900.1 | - | 1 | 175 |