Gene Transcripts

Gene Symbol
Dclk1
Entrez Gene ID
13175
Gene Name
doublecortin-like kinase 1
Gene Aliases
1700113D08Rik, 2810480F11Rik, AI836758, Click-I, Cpg16, Dcamkl1, Dcl, Dclk, mKIAA0369
Chromosome Location
Chr. 3: 55242369 - 55539068 on Build GRCm38
UniGene ID
Mm.393242
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001111051.1 NP_001104521.1 13 4957
NM_001111052.1 NP_001104522.1 14 5031
NM_001195538.1 NP_001182467.1 17 5818
NM_001195539.1 NP_001182468.1 13 5005
NM_019978.3 NP_064362.1 17 5866
GenBank mRNA AK034632.1 - 1 2832

View Full Product Details