Gene Transcripts
- Gene Symbol
- TMEM68
- Entrez Gene ID
- 137695
- Gene Name
- transmembrane protein 68
- Gene Aliases
- Chromosome Location
- Chr. 8: 55738744 - 55773407 on Build GRCh38
- UniGene ID
- Hs.420076
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001286657.1 |
NP_001273586.1
|
3, 4 |
616 |
|
NM_001286661.1 |
NP_001273590.1
|
2, 3 |
316 |
|
NM_152417.2 |
NP_689630.1
|
4, 5 |
755 |
|
XM_005251150.1 |
XP_005251207.1
|
4, 5 |
744 |
|
XM_011517454.1 |
XP_011515756.1
|
4, 5 |
768 |
|
XM_011517455.2 |
XP_011515757.1
|
4, 5 |
744 |
|
XM_017013040.1 |
XP_016868529.1
|
3, 4 |
328 |
|
GenBank mRNA |
AK056932.1 |
- |
4, 5 |
674 |
|
AL832935.1 |
- |
2, 3 |
225 |
|
CN272366.1 |
- |
2, 3 |
243 |
|
View Full Product Details