Gene Transcripts
- Gene Symbol
- Rps8
- Entrez Gene ID
- 20116
- Gene Name
- ribosomal protein S8
- Gene Aliases
- Chromosome Location
- Chr. 4: 117153836 - 117156132 on Build GRCm38
- UniGene ID
- Mm.260904
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_009098.2 |
NP_033124.1
|
3, 4 |
286 |
|
GenBank mRNA |
AK009023.1 |
- |
3, 4 |
239 |
|
AK010306.1 |
- |
3, 4 |
231 |
|
AK010650.1 |
- |
3, 4 |
286 |
|
AK011048.1 |
- |
3, 4 |
255 |
|
AK011058.1 |
- |
3, 4 |
239 |
|
AK011642.1 |
- |
3, 4 |
231 |
|
AK012435.1 |
- |
3, 4 |
239 |
|
AK012562.1 |
- |
3, 4 |
239 |
|
AK012665.1 |
- |
3, 4 |
258 |
|
AK019223.1 |
- |
3, 4 |
239 |
|
AK145478.1 |
- |
2, 3 |
702 |
|
AK146303.1 |
- |
3, 4 |
239 |
|
AK146806.1 |
- |
3, 4 |
239 |
|
AK151544.1 |
- |
3, 4 |
239 |
|
AK166848.1 |
- |
3, 4 |
239 |
|
AK168609.1 |
- |
3, 4 |
239 |
|
BC027217.1 |
- |
2, 3 |
218 |
4205924 |
BC051446.1 |
- |
2, 3 |
235 |
30054488 |
BC081464.1 |
- |
3, 4 |
246 |
|
BC081465.1 |
- |
2, 3 |
218 |
6823252 |
BC086899.1 |
- |
3, 4 |
238 |
6748012 |
BC100594.1 |
- |
3, 4 |
238 |
6750930 |
X73829.1 |
- |
2, 3 |
221 |
|
- Gene Symbol
- Gm15501
- Entrez Gene ID
- 100040298
- Gene Name
- ribosomal protein S8 pseudogene
- Gene Aliases
- ENSMUSG00000070446, OTTMUSG00000022847
- Chromosome Location
- Chr. 7: 93178980 - 93179684 on Build GRCm38
- UniGene ID
- Mm.260904
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
CJ089270.1 |
- |
1 |
239 |
|
DV661469.1 |
- |
|
401 |
|
View Full Product Details