Gene Transcripts

Gene Symbol
Ribc2
Entrez Gene ID
67747
Gene Name
RIB43A domain with coiled-coils 2
Gene Aliases
4930579A10Rik, Trib
Chromosome Location
Chr. 15: 85132060 - 85144569 on Build GRCm38
UniGene ID
Mm.158173
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026357.2 NP_080633.2 3 495
GenBank mRNA AK006801.1 - 3 492
AK016311.1 - 3 496
BC112373.1 - 3 420 40056860

View Full Product Details