Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027102.3 | NP_081378.1 | 4 | 682 | |
GenBank mRNA | AF361882.1 | - | 4 | 667 | |
AK009223.1 | - | 4 | 658 | ||
AK156399.1 | - | 4 | 680 | ||
BC019460.1 | - | 4 | 666 | 4188050 | |
BC049249.1 | - | 4 | 662 | 5039892 | |
BG077536.2 | - | 2 | 116 |