Gene Transcripts
- Gene Symbol
- Kansl2
- Entrez Gene ID
- 69612
- Gene Name
- KAT8 regulatory NSL complex subunit 2
- Gene Aliases
- 2310037I24Rik, B930009D17
- Chromosome Location
- Chr. 15: 98517658 - 98534732 on Build GRCm38
- UniGene ID
- Mm.229107
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001289437.1 |
NP_001276366.1
|
7, 8 |
1349 |
|
NM_001289438.1 |
NP_001276367.1
|
7, 8 |
1441 |
|
NM_001289439.1 |
NP_001276368.1
|
6, 7 |
1234 |
|
NM_001289440.1 |
NP_001276369.1
|
6, 7 |
895 |
|
NM_133714.5 |
NP_598475.2
|
7, 8 |
1349 |
|
GenBank mRNA |
AK046649.1 |
- |
7, 8 |
1319 |
|
AK046983.1 |
- |
3, 4 |
288 |
|
AK047269.1 |
- |
3, 4 |
287 |
|
AK089025.1 |
- |
6, 7 |
1223 |
|
AK150748.1 |
- |
7, 8 |
1343 |
|
AK151604.1 |
- |
7, 8 |
1344 |
|
AK152604.1 |
- |
7, 8 |
1343 |
|
AK153007.1 |
- |
7, 8 |
1343 |
|
AK162714.1 |
- |
7, 8 |
1322 |
|
AK167429.1 |
- |
7, 8 |
1324 |
|
AK168499.1 |
- |
6, 7 |
1212 |
|
AK183493.1 |
- |
3, 4 |
290 |
|
AK188822.1 |
- |
3, 4 |
289 |
|
AK205712.1 |
- |
3, 4 |
290 |
|
AK218567.1 |
- |
3, 4 |
308 |
|
BC003301.1 |
- |
7, 8 |
1325 |
3584464 |
BC011085.1 |
- |
6, 7 |
890 |
|
BC083325.1 |
- |
7, 8 |
1065 |
6828438 |
BC092275.1 |
- |
5, 6 |
661 |
3492339 |
BC125631.1 |
- |
7, 8 |
1301 |
40130146 |
BC125633.1 |
- |
7, 8 |
1301 |
40130148 |
BC145047.1 |
- |
7, 8 |
1393 |
|
View Full Product Details