Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033371.2 | NP_203535.1 | 11 | 2094 | |
GenBank mRNA | AK013817.1 | - | 4 | 1088 | |
AK134809.1 | - | 3 | 1060 | ||
AK150235.1 | - | 8 | 890 | ||
AK154865.1 | - | 11 | 2079 | ||
AK171452.1 | - | 11 | 2079 | ||
AK191582.1 | - | 1 | 94 | ||
AK220248.1 | - | 9 | 1892 | ||
AY010723.1 | - | 10 | 1289 | ||
BC033318.1 | - | 8 | 1452 | ||
BC044775.1 | - | 9 | 1849 | ||
BC049958.1 | - | 9 | 1849 | ||
BC057450.1 | - | 10 | 1711 | 3966334 | |
BC096620.1 | - | 11 | 2094 | 6390076 |