Gene Transcripts
- Gene Symbol
- Camkmt
- Entrez Gene ID
- 73582
- Gene Name
- calmodulin-lysine N-methyltransferase
- Gene Aliases
- 1700106N22Rik, AI480743, AV099781, CLNMT, CaM KMT
- Chromosome Location
- Chr. 17: 85090592 - 85458589 on Build GRCm38
- UniGene ID
- Mm.260753
- Species
- Mus musculus
- Gene Symbol
- Gnao1
- Entrez Gene ID
- 14681
- Gene Name
- guanine nucleotide binding protein, alpha O
- Gene Aliases
- AW050213, Galphao, Gnao, alphaO
- Chromosome Location
- Chr. 8: 93810561 - 93969388 on Build GRCm38
- UniGene ID
- Mm.251445
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK147783.1 |
- |
|
349 |
|
View Full Product Details