Gene Transcripts

Gene Symbol
Ppp1r3g
Entrez Gene ID
76487
Gene Name
protein phosphatase 1, regulatory (inhibitor) subunit 3G
Gene Aliases
1600032L17Rik
Chromosome Location
Chr. 13: 35958799 - 35970388 on Build GRCm38
UniGene ID
Mm.44745
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029628.1 NP_083904.1 2 1488
GenBank mRNA AK005570.1 - 1 798
AK050457.1 - 1 194
AK079637.1 - 2 1462

View Full Product Details