Gene Transcripts

Gene Symbol
ZNF674
Entrez Gene ID
641339
Gene Name
zinc finger protein 674
Gene Aliases
MRX92, ZNF673B
Chromosome Location
Chr. X: 46497725 - 46545459 on Build GRCh38
UniGene ID
Hs.636105
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039891.2 NP_001034980.1 6 554
NM_001146291.1 NP_001139763.1 6 536
NM_001190417.1 NP_001177346.1 6 539
XM_011543941.2 XP_011542243.1 6 551
XM_011543943.2 XP_011542245.1 6 553
XM_011543944.2 XP_011542246.1 4 361
XM_011543945.2 XP_011542247.1 3 310
XM_017029728.1 XP_016885217.1 7 706
XM_017029729.1 XP_016885218.1 7 728
XM_017029730.1 XP_016885219.1 7 725
XM_017029731.1 XP_016885220.1 3 310
XM_017029732.1 XP_016885221.1 7 703
XM_017029733.1 XP_016885222.1 7 705
GenBank mRNA AK295054.1 - 6 468
AY971607.1 - 6 538
DC317540.1 - 6 523

Gene Symbol
KRBOX4
Entrez Gene ID
55634
Gene Name
KRAB box domain containing 4
Gene Aliases
ZNF673
Chromosome Location
Chr. X: 46447189 - 46474639 on Build GRCh38
UniGene ID
Hs.632800
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001129898.1 NP_001123370.1 6 626
NM_001129899.1 NP_001123371.1 7 670
NM_001129900.1 NP_001123372.1 7 655
NM_017776.2 NP_060246.2 6 611
GenBank mRNA AK000351.1 - 6 476
AK096413.1 - 6 518
AK097159.1 - 1 1500
AK289703.1 - 6 508
BC012569.1 - 6 478 4283420
DA631861.1 - 7 572

View Full Product Details