Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006271.1 | NP_006262.1 | 3 | 301 | |
GenBank mRNA | AB451316.1 | - | 2 | 188 | |
AB451446.1 | - | 2 | 188 | ||
AK126904.1 | - | 2 | 2326 | ||
AK312152.1 | - | 3 | 258 | ||
BC014392.1 | - | 3 | 263 | 3543900 | |
BC072006.1 | - | 3 | 422 | ||
BC094843.1 | - | 3 | 424 | ||
BT006938.1 | - | 2 | 188 | ||
KU178317.1 | - | 2 | 188 | ||
KU178318.1 | - | 2 | 374 | ||
X58079.1 | - | 3 | 301 |