Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001111097.2 | NP_001104567.1 | 4 | 413 | |
NM_002350.3 | NP_002341.1 | 4 | 476 | ||
XM_011517529.2 | XP_011515831.1 | 3 | 2861 | ||
XM_017013415.1 | XP_016868904.1 | 3 | 2861 | ||
XM_017013416.1 | XP_016868905.1 | 4 | 152 | ||
GenBank mRNA | AK290494.1 | - | 4 | 386 | |
AK298677.1 | - | 2 | 64 | ||
BC059394.1 | - | 4 | 405 | ||
BC068551.1 | - | 4 | 405 | ||
BC075001.2 | - | 3 | 219 | 30915399 | |
BC075002.2 | - | 3 | 219 | 30915611 | |
BC126456.1 | - | 5 | 313 | 8992172 | |
BC126458.1 | - | 5 | 360 | 8992174 | |
CB990993.1 | - | 4 | 438 | ||
M16038.1 | - | 4 | 491 | ||
M79321.1 | - | 3 | 155 |