gRNA Design Details
- Target DNA Sequence
- GATCTCTCCAACAACCGGAT
- PAM Sequence
- TGG
- Target locus
-
Chr.8: 37829262 - 37829284 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
89.04/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ADGRA2
- Entrez Gene ID
-
25960
- Gene Name
- adhesion G protein-coupled receptor A2
- Gene Aliases
-
GPR124, TEM5
- Chromosome Location
-
Chr.8: 37796883 - 37843986 on GRCh38
- UniGene ID
- Hs.274136
- Species
-
Human