gRNA Design Details

Target DNA Sequence
GTTCCTGTTCACGGCCACGT
PAM Sequence
GGG
Target locus
Chr.10: 133128617 - 133128639 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 95.36/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
ADGRA1
Entrez Gene ID
84435
Gene Name
adhesion G protein-coupled receptor A1
Gene Aliases
GPR123
Chromosome Location
Chr.10: 133070929 - 133131675 on GRCh38
UniGene ID
Hs.435183
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq XM_005252695.1 XP_005252752.1 16