gRNA Design Details
- Target DNA Sequence
- GCGGATGGATAGCGGGCAGC
- PAM Sequence
- CGG
- Target locus
-
Chr.8: 37797372 - 37797394 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
87.54/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ADGRA2
- Entrez Gene ID
-
25960
- Gene Name
- adhesion G protein-coupled receptor A2
- Gene Aliases
-
GPR124, TEM5
- Chromosome Location
-
Chr.8: 37796883 - 37843986 on GRCh38
- UniGene ID
- Hs.274136
- Species
-
Human