gRNA Design Details
- Target DNA Sequence
- ATCGGATCTATTCGGACCCT
- PAM Sequence
- AGG
- Target locus
-
Chr.11: 105969018 - 105969040 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
87.95/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Ace
- Entrez Gene ID
-
11421
- Gene Name
- angiotensin I converting enzyme (peptidyl-dipeptidase A) 1
- Gene Aliases
-
AW208573, CD143
- Chromosome Location
-
Chr.11: 105967945 - 105989964 on GRCm38
- UniGene ID
- Mm.754
- Species
-
Mouse