gRNA Design Details
- Target DNA Sequence
- AGTCACACCGATCCACAGCA
- PAM Sequence
- TGG
- Target locus
-
Chr.7: 139852546 - 139852568 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
86.29/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Adgra1
- Entrez Gene ID
-
52389
- Gene Name
- adhesion G protein-coupled receptor A1
- Gene Aliases
-
2900059M17Rik, D7Ertd680e, Gpr123
- Chromosome Location
-
Chr.7: 139834084 - 139878088 on GRCm38
- UniGene ID
- Mm.39863
- Species
-
Mouse