gRNA Design Details
- Target DNA Sequence
- ACGAGTATGCCGAGGCCAAC
- PAM Sequence
- TGG
- Target locus
-
Chr.17: 63485308 - 63485330 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
87/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ACE
- Entrez Gene ID
-
1636
- Gene Name
- angiotensin I converting enzyme
- Gene Aliases
-
ACE1, CD143, DCP, DCP1, ICH, MVCD3
- Chromosome Location
-
Chr.17: 63477061 - 63498380 on GRCh38
- UniGene ID
- Hs.298469
- Species
-
Human