Gene Transcripts
- Gene Symbol
- Cops8
- Entrez Gene ID
- 108679
- Gene Name
- COP9 (constitutive photomorphogenic) homolog, subunit 8 (Arabidopsis thaliana)
- Gene Aliases
- 9430009J09Rik, AA408242, Csn8, Sgn8
- Chromosome Location
- Chr. 1: 90603425 - 90613341 on Build GRCm38
- UniGene ID
- Mm.119177
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_133805.3 |
NP_598566.3
|
4, 5 |
444 |
|
GenBank mRNA |
AF482000.1 |
- |
4, 5 |
905 |
|
AK079111.1 |
- |
4, 5 |
378 |
|
AK079522.1 |
- |
4, 5 |
392 |
|
AK088323.1 |
- |
4, 5 |
364 |
|
AK090013.1 |
- |
4, 5 |
373 |
|
AK142989.1 |
- |
3, 4 |
1413 |
|
AK145899.1 |
- |
4, 5 |
371 |
|
AK162502.1 |
- |
4, 5 |
379 |
|
BC017690.1 |
- |
4, 5 |
412 |
4238947 |
BC021488.1 |
- |
4, 5 |
403 |
5344864 |
BC024421.1 |
- |
4, 5 |
397 |
4987584 |
View Full Product Details