Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_183020.1 | NP_898841.1 | 4, 5 | 572 | |
GenBank mRNA | AK084800.1 | - | 4, 5 | 644 | |
AK158626.1 | - | 4, 5 | 259 | ||
AK168745.1 | - | 4, 5 | 289 | ||
BC054483.1 | - | 4, 5 | 572 | 6391591 |