Gene Transcripts

Gene Symbol
Dll4
Entrez Gene ID
54485
Gene Name
delta-like 4 (Drosophila)
Gene Aliases
Delta4
Chromosome Location
Chr. 2: 119325784 - 119335666 on Build GRCm38
UniGene ID
Mm.143719
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019454.3 NP_062327.2 9, 10 2295
GenBank mRNA AB043893.1 - 9, 10 1938
AF253469.1 - 9, 10 2250
AK004739.1 - 9, 10 2263
AK031699.1 - 9, 10 2265
AK052322.1 - 9, 10 2297
AK165517.1 - 9, 10 2263
AK171717.1 - 9, 10 2262
BC042497.1 - 9, 10 2250 4017786
BC049130.1 - 9, 10 2255 6825525

View Full Product Details