Gene Transcripts
- Gene Symbol
- LOC100287290
- Entrez Gene ID
- 100287290
- Gene Name
- cytokine receptor CRL2
- Gene Aliases
- Chromosome Location
- Chr. 3: 158732441 - 158784552 on Build GRCh38
- UniGene ID
- Hs.616897
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK093960.1 |
- |
1 |
1550 |
|
View Full Product Details