Gene Transcripts
- Gene Symbol
- SEMA6A
- Entrez Gene ID
- 57556
- Gene Name
- semaphorin 6A
- Gene Aliases
- HT018, SEMA, SEMA6A1, SEMAQ, VIA
- Chromosome Location
- Chr. 5: 116443555 - 116574926 on Build GRCh38
- UniGene ID
- Hs.156967
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK096337.1 |
- |
1 |
1311 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
U06699.1 |
- |
1 |
369 |
|
Chr. 5 - 116466658 - 116468601 [-] |
View Full Product Details