Gene Transcripts

Gene Symbol
Mettl20
Entrez Gene ID
320204
Gene Name
methyltransferase like 20
Gene Aliases
4833442J19Rik, AI256744
Chromosome Location
Chr. 6: 149141513 - 149151171 on Build GRCm38
UniGene ID
Mm.274099
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252094.1 NP_001239023.1 4 756
NM_001252095.1 NP_001239024.1 4 737
NM_001252096.1 NP_001239025.1 3 646
NM_001252097.1 NP_001239026.1 2 218
NM_177101.5 NP_796075.2 3 627
GenBank mRNA AK132096.1 - 4 624
BC046911.1 - 2 157 6491688
BC048711.1 - 3 519 6328428
BC065807.1 - 3 543 3373106
DN174886.1 - 3 520

View Full Product Details