Gene Transcripts

Gene Symbol
P2rx4
Entrez Gene ID
29659
Gene Name
purinergic receptor P2X, ligand-gated ion channel 4
Gene Aliases
Chromosome Location
Chr. 12: 39308022 - 39325673 on Build Rnor_6.0
UniGene ID
Rn.7176
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031594.1 NP_113782.1 3 419
GenBank mRNA BC078792.1 - 3 360
U32497.1 - 3 396
X87763.1 - 3 419
X93565.1 - 3 506

View Full Product Details