Gene Transcripts

Gene Symbol
MID1
Entrez Gene ID
4281
Gene Name
midline 1
Gene Aliases
BBBG1, FXY, GBBB1, MIDIN, OGS1, OS, OSX, RNF59, TRIM18, XPRF, ZNFXY
Chromosome Location
Chr. X: 10445310 - 11111264 on Build GRCh38
UniGene ID
Hs.27695
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000381.3 NP_000372.1 8 1708
NM_001098624.2 NP_001092094.1 8 1638
NM_001193277.1 NP_001180206.1 8 1497
NM_033289.1 NP_150631.1 8 1594
NM_033290.3 NP_150632.1 8 1500
XM_005274536.1 XP_005274593.1 9 1673
XM_005274537.1 XP_005274594.1 9 1650
XM_006724492.3 XP_006724555.1 9 2272
XM_011545525.2 XP_011543827.1 11 1974
XM_011545526.2 XP_011543828.1 11 2009
XM_011545527.2 XP_011543829.1 9 1919
XM_017029534.1 XP_016885023.1 9 1839
XM_017029535.1 XP_016885024.1 11 1860
XM_017029536.1 XP_016885025.1 10 1936
XM_017029537.1 XP_016885026.1 10 1707
XM_017029538.1 XP_016885027.1 8 1406
XM_017029539.1 XP_016885028.1 8 1568
XM_017029540.1 XP_016885029.1 10 1821
GenBank mRNA AF035360.1 - 8 1394
AF041206.1 - 8 1588
AF041208.1 - 8 1708
AF041210.1 - 7 1197
AF230976.1 - 8 1493
AF230977.1 - 8 1493
AF269101.1 - 9 1616
AK025187.1 - 7 678
AK226104.1 - 8 1412
AK291215.1 - 8 1590
AK297205.1 - 9 1504
AK299061.1 - 8 1562
AK315095.1 - 8 1539
AY539962.1 - 7 690
AY539983.1 - 7 733
AY539997.1 - 6 607
AY540009.1 - 4 377
AY540010.1 - 4 482
AY540011.1 - 3 439
BC053626.1 - 8 1565 6163881
Y13667.1 - 8 1493

View Full Product Details