Gene Transcripts
- Gene Symbol
- INHBA
- Entrez Gene ID
- 3624
- Gene Name
- inhibin beta A subunit
- Gene Aliases
- EDF, FRP
- Chromosome Location
- Chr. 7: 41685101 - 41710532 on Build GRCh38
- UniGene ID
- Hs.28792
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
J03634.1 |
- |
2 |
1564 |
|
View Full Product Details