Gene Transcripts

Gene Symbol
ATP2B4
Entrez Gene ID
493
Gene Name
ATPase plasma membrane Ca2+ transporting 4
Gene Aliases
ATP2B2, MXRA1, PMCA4, PMCA4b, PMCA4x
Chromosome Location
Chr. 1: 203626787 - 203744081 on Build GRCh38
UniGene ID
Hs.343522
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001396.2 NP_001001396.1 3 1203
NM_001684.4 NP_001675.3 3 1203
GenBank mRNA AK292458.1 - 2 542
BC140774.1 - 3 875 9021642
BC150293.1 - 2 560 8860318
BX537444.1 - 3 1430
CR749393.1 - 3 1133
CR936708.1 - 3 1157
M25874.1 - 2 703
M83363.1 - 2 703

View Full Product Details