Gene Transcripts

Gene Symbol
SP140
Entrez Gene ID
11262
Gene Name
SP140 nuclear body protein
Gene Aliases
LYSP100, LYSP100-A, LYSP100-B
Chromosome Location
Chr. 2: 230202924 - 230316571 on Build GRCh38
UniGene ID
Hs.632549
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005176.2 NP_001005176.1 2 204
NM_001278451.1 NP_001265380.1 2 204
NM_001278452.1 NP_001265381.1 2 204
NM_001278453.1 NP_001265382.1 2 204
NM_007237.4 NP_009168.4 2 204
XM_005246252.2 XP_005246309.1 2 168
XM_005246253.2 XP_005246310.1 2 168
XM_005246254.2 XP_005246311.1 2 168
XM_005246255.2 XP_005246312.1 2 168
XM_005246256.2 XP_005246313.1 2 168
XM_006712223.3 XP_006712286.1 2 168
XM_011510515.1 XP_011508817.1 2 168
XM_011510516.1 XP_011508818.1 2 168
XM_011510517.2 XP_011508819.1 4 1032
XM_011510518.2 XP_011508820.1 3 304
XM_011510519.1 XP_011508821.1 2 168
XM_011510520.1 XP_011508822.1 2 168
XM_017003239.1 XP_016858728.1 2 168
XM_017003240.1 XP_016858729.1 2 168
XM_017003241.1 XP_016858730.1 2 168
XM_017003242.1 XP_016858731.1 2 168
XM_017003243.1 XP_016858732.1 2 168
XM_017003244.1 XP_016858733.1 3 304
XM_017003245.1 XP_016858734.1 2 168
XM_017003246.1 XP_016858735.1 2 168
XM_017003247.1 XP_016858736.1 2 168
XM_017003248.1 XP_016858737.1 2 168
XM_017003249.1 XP_016858738.1 2 168
XM_017003250.1 XP_016858739.1 2 168
XM_017003251.1 XP_016858740.1 3 311
XM_017003252.1 XP_016858741.1 2 168
GenBank mRNA AK301267.1 - 2 204
AK301317.1 - 2 205
BC038091.1 - 2 175
BC054890.1 - 2 174
BC070160.1 - 2 170
BC105743.1 - 2 187 40035747
BC105744.1 - 2 189 40035746
BC105960.1 - 2 189 40035752
DC391925.1 - 2 204
U36499.1 - 2 205
U36500.1 - 2 205
U63420.1 - 2 195

View Full Product Details