Gene Transcripts
- Gene Symbol
- ARSF
- Entrez Gene ID
- 416
- Gene Name
- arylsulfatase F
- Gene Aliases
- ASF
- Chromosome Location
- Chr. X: 3040234 - 3112729 on Build GRCh38
- UniGene ID
- Hs.101674
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
X97868.1 |
- |
4 |
167 |
|
View Full Product Details