Gene Transcripts
- Gene Symbol
- Mnx1
- Entrez Gene ID
- 15285
- Gene Name
- motor neuron and pancreas homeobox 1
- Gene Aliases
- HB9, Hlxb9, MNR2
- Chromosome Location
- Chr. 5: 29473823 - 29478470 on Build GRCm38
- UniGene ID
- Mm.103760
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF153046.1 |
- |
|
1865 |
|
View Full Product Details