Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001277216.1 | NP_001264145.1 | 12 | 1905 | |
NM_001277217.1 | NP_001264146.1 | 12 | 1931 | ||
NM_001277218.1 | NP_001264147.1 | 11 | 1944 | ||
NM_001277220.1 | NP_001264149.1 | 12 | 2079 | ||
NM_007560.4 | NP_031586.1 | 11 | 2098 | ||
GenBank mRNA | AK086130.1 | - | 12 | 1891 | |
AK140158.1 | - | 12 | 2003 | ||
AK162844.1 | - | 11 | 1779 | ||
BC065106.1 | - | 11 | 1879 | 30357130 | |
BC065143.1 | - | 11 | 2085 | 30629345 | |
BC138638.1 | - | 11 | 1825 | 8861658 | |
BC138640.1 | - | 11 | 1825 | 8861660 | |
Z23143.1 | - | 12 | 1923 |