Gene Transcripts
- Gene Symbol
- Abl1
- Entrez Gene ID
- 11350
- Gene Name
- c-abl oncogene 1, non-receptor tyrosine kinase
- Gene Aliases
- AI325092, Abl, E430008G22Rik, c-Abl
- Chromosome Location
- Chr. 2: 31688252 - 31807093 on Build GRCm38
- UniGene ID
- Mm.1318
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001112703.2 |
NP_001106174.1
|
2, 3 |
1577 |
|
NM_001283045.1 |
NP_001269974.1
|
2, 3 |
454 |
|
NM_001283046.1 |
NP_001269975.1
|
3, 4 |
1782 |
|
NM_001283047.1 |
NP_001269976.1
|
3, 4 |
1721 |
|
NM_009594.4 |
NP_033724.2
|
2, 3 |
348 |
|
GenBank mRNA |
AK088252.1 |
- |
3, 4 |
1707 |
|
AK090095.1 |
- |
2, 3 |
306 |
|
AK166028.1 |
- |
2, 3 |
1566 |
|
AK166210.1 |
- |
3, 4 |
1816 |
|
AK192334.1 |
- |
|
407 |
|
AK194745.1 |
- |
|
285 |
|
BC059260.1 |
- |
2, 3 |
375 |
6408990 |
BC103770.1 |
- |
2, 3 |
801 |
30848553 |
BY152805.1 |
- |
2, 3 |
347 |
|
CJ099623.1 |
- |
2, 3 |
299 |
|
CJ128107.1 |
- |
2, 3 |
343 |
|
CN663033.1 |
- |
1, 2 |
69 |
|
J02995.1 |
- |
2, 3 |
301 |
|
K03228.1 |
- |
2, 3 |
343 |
|
M12263.1 |
- |
3, 4 |
708 |
|
M12264.1 |
- |
2, 3 |
454 |
|
M12265.1 |
- |
2, 3 |
503 |
|
View Full Product Details