Gene Transcripts
- Gene Symbol
- Tecr
- Entrez Gene ID
- 191576
- Gene Name
- trans-2,3-enoyl-CoA reductase
- Gene Aliases
- Gpsn2, SC2
- Chromosome Location
- Chr. 19: 24705478 - 24732029 on Build Rnor_6.0
- UniGene ID
- Rn.107358
- Species
- Rattus norvegicus
- Gene Symbol
- RGD1560015
- Entrez Gene ID
- 499018
- Gene Name
- similar to glycoprotein, synaptic 2
- Gene Aliases
- Chromosome Location
- Chr. 1: 47587022 - 47588091 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_574311.6 |
XP_574311.1
|
1 |
633 |
|
View Full Product Details