Gene Transcripts

Gene Symbol
N6AMT1
Entrez Gene ID
29104
Gene Name
N-6 adenine-specific DNA methyltransferase 1 (putative)
Gene Aliases
C21orf127, HEMK2, MTQ2, N6AMT, PRED28, m.HsaHemK2P
Chromosome Location
Chr. 21: 28571095 - 28885373 on Build GRCh38
UniGene ID
Hs.163846
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013240.5 NP_037372.4 2 199
NM_182749.4 NP_877426.4 2 199
NR_047510.2 2 199
GenBank mRNA AF139682.1 - 2 200
AK225515.1 - 2 193
AK314100.1 - 2 191
BC011554.1 - 2 180 4640080
BP317563.1 - 2 196

View Full Product Details