Gene Transcripts

Gene Symbol
POM121C
Entrez Gene ID
100101267
Gene Name
POM121 transmembrane nucleoporin C
Gene Aliases
POM121-2
Chromosome Location
Chr. 7: 75416781 - 75486288 on Build GRCh38
UniGene ID
Hs.712563
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK125865.1 - 1 3447
AK127026.1 - 2 3803
AL050158.1 - 1 1283
BX649044.1 - 2 2158

View Full Product Details