Gene Transcripts

Gene Symbol
OBSCN
Entrez Gene ID
84033
Gene Name
obscurin, cytoskeletal calmodulin and titin-interacting RhoGEF
Gene Aliases
ARHGEF30, UNC89
Chromosome Location
Chr. 1: 228208013 - 228381431 on Build GRCh38
UniGene ID
Hs.650039
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098623.2 NP_001092093.2 94 22033
NM_001271223.2 NP_001258152.2 105 24904
XM_005273287.4 XP_005273344.1 106 25290
XM_005273291.4 XP_005273348.1 106 25254
XM_005273298.4 XP_005273355.1 105 24981
XM_005273307.4 XP_005273364.1 101 24168
XM_006711822.3 XP_006711885.1 104 24714
XM_006711823.3 XP_006711886.1 104 24708
XM_011544297.2 XP_011542599.1 98 23124
XM_011544299.2 XP_011542601.1 97 22848
XM_017002443.1 XP_016857932.1 106 25335
XM_017002444.1 XP_016857933.1 106 25298
XM_017002445.1 XP_016857934.1 106 25332
XM_017002446.1 XP_016857935.1 106 25332
XM_017002447.1 XP_016857936.1 106 25332
XM_017002448.1 XP_016857937.1 106 25185
XM_017002449.1 XP_016857938.1 105 25072
XM_017002450.1 XP_016857939.1 105 25072
XM_017002451.1 XP_016857940.1 105 25072
XM_017002452.1 XP_016857941.1 105 25072
XM_017002453.1 XP_016857942.1 105 25059
XM_017002454.1 XP_016857943.1 105 25059
XM_017002455.1 XP_016857944.1 105 25059
XM_017002456.1 XP_016857945.1 105 25059
XM_017002457.1 XP_016857946.1 105 25059
XM_017002458.1 XP_016857947.1 104 24808
XM_017002459.1 XP_016857948.1 104 24808
XM_017002460.1 XP_016857949.1 104 24808
XM_017002461.1 XP_016857950.1 104 24795
XM_017002462.1 XP_016857951.1 104 24783
XM_017002463.1 XP_016857952.1 104 24762
XM_017002464.1 XP_016857953.1 104 24711
XM_017002465.1 XP_016857954.1 103 24544
XM_017002466.1 XP_016857955.1 103 24544
XM_017002467.1 XP_016857956.1 103 24544
XM_017002468.1 XP_016857957.1 102 24280
XM_017002469.1 XP_016857958.1 100 23752
XM_017002470.1 XP_016857959.1 95 22433
GenBank mRNA AB046859.2 - 34 5808
AM231061.1 - 20 3935
DQ400343.1 - 20 3935

View Full Product Details