Gene Transcripts

Gene Symbol
OBSCN
Entrez Gene ID
84033
Gene Name
obscurin, cytoskeletal calmodulin and titin-interacting RhoGEF
Gene Aliases
ARHGEF30, UNC89
Chromosome Location
Chr. 1: 228208013 - 228381431 on Build GRCh38
UniGene ID
Hs.650039
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098623.2 NP_001092093.2 94 22013
NM_001271223.2 NP_001258152.2 105 24884
XM_005273287.4 XP_005273344.1 106 25270
XM_005273291.4 XP_005273348.1 106 25234
XM_005273298.4 XP_005273355.1 105 24961
XM_005273307.4 XP_005273364.1 101 24148
XM_006711822.3 XP_006711885.1 104 24694
XM_006711823.3 XP_006711886.1 104 24688
XM_011544297.2 XP_011542599.1 98 23104
XM_011544299.2 XP_011542601.1 97 22828
XM_017002443.1 XP_016857932.1 106 25315
XM_017002444.1 XP_016857933.1 106 25278
XM_017002445.1 XP_016857934.1 106 25312
XM_017002446.1 XP_016857935.1 106 25312
XM_017002447.1 XP_016857936.1 106 25312
XM_017002448.1 XP_016857937.1 106 25165
XM_017002449.1 XP_016857938.1 105 25052
XM_017002450.1 XP_016857939.1 105 25052
XM_017002451.1 XP_016857940.1 105 25052
XM_017002452.1 XP_016857941.1 105 25052
XM_017002453.1 XP_016857942.1 105 25039
XM_017002454.1 XP_016857943.1 105 25039
XM_017002455.1 XP_016857944.1 105 25039
XM_017002456.1 XP_016857945.1 105 25039
XM_017002457.1 XP_016857946.1 105 25039
XM_017002458.1 XP_016857947.1 104 24788
XM_017002459.1 XP_016857948.1 104 24788
XM_017002460.1 XP_016857949.1 104 24788
XM_017002461.1 XP_016857950.1 104 24775
XM_017002462.1 XP_016857951.1 104 24763
XM_017002463.1 XP_016857952.1 104 24742
XM_017002464.1 XP_016857953.1 104 24691
XM_017002465.1 XP_016857954.1 103 24524
XM_017002466.1 XP_016857955.1 103 24524
XM_017002467.1 XP_016857956.1 103 24524
XM_017002468.1 XP_016857957.1 102 24260
XM_017002469.1 XP_016857958.1 100 23732
XM_017002470.1 XP_016857959.1 95 22413
GenBank mRNA AB046859.2 - 34 5788
AM231061.1 - 20 3915
DQ400343.1 - 20 3915

View Full Product Details