Gene Transcripts
- Gene Symbol
- HYDIN
- Entrez Gene ID
- 54768
- Gene Name
- HYDIN, axonemal central pair apparatus protein
- Gene Aliases
- CILD5, HYDIN1, HYDIN2, PPP1R31
- Chromosome Location
- Chr. 16: 70802084 - 71230722 on Build GRCh38
- UniGene ID
- Hs.351914
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001270974.2 |
NP_001257903.1
|
53, 54 |
9230 |
|
XM_006721206.2 |
XP_006721269.1
|
53, 54 |
9124 |
|
XM_011523146.2 |
XP_011521448.1
|
54, 55 |
9259 |
|
XM_011523147.1 |
XP_011521449.1
|
54, 55 |
9226 |
|
XM_011523148.1 |
XP_011521450.1
|
54, 55 |
9299 |
|
XM_011523151.1 |
XP_011521453.1
|
53, 54 |
9156 |
|
XM_011523152.1 |
XP_011521454.1
|
16, 17 |
2983 |
|
XM_011523155.2 |
XP_011521457.1
|
9, 10 |
1984 |
|
XM_017023346.1 |
XP_016878835.1
|
53, 54 |
9402 |
|
XM_017023347.1 |
XP_016878836.1
|
41, 42 |
7641 |
|
XM_017023348.1 |
XP_016878837.1
|
41, 42 |
7381 |
|
- Gene Symbol
- HYDIN2
- Entrez Gene ID
- 100288805
- Gene Name
- HYDIN2, axonemal central pair apparatus protein (pseudogene)
- Gene Aliases
- HYDINP1
- Chromosome Location
- Chr. 1: 146547489 - 146822034 on Build GRCh38
- UniGene ID
- Hs.648712
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_103556.1 |
|
44, 45 |
7752 |
|
GenBank mRNA |
AB058767.1 |
- |
18, 19 |
3484 |
|
View Full Product Details