Gene Transcripts
- Gene Symbol
- GUSBP1
- Entrez Gene ID
- 728411
- Gene Name
- glucuronidase, beta pseudogene 1
- Gene Aliases
- Chromosome Location
- Chr. 5: 21459480 - 21589372 on Build GRCh38
- UniGene ID
- Hs.88181
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027026.1 |
|
5 |
1540 |
|
GenBank mRNA |
AK095656.1 |
- |
5 |
1464 |
|
View Full Product Details